Learn More
Medchemexpress LLC Propacetamol (hydrochloride) | 66532-86-3 | MFCD00941392 | 300.79 | C14H21ClN2O3 | 100 MG
SDP
Catalog No. 5000243259
Encompass_Preferred
Propacetamol (hydrochloride) is the hydrochloride salt of propacetamol, a water-soluble prodrug of paracetamol (acetaminophen) used as an analgesic in research settings. Supplied for biochemical and in vivo studies, it is provided in solid and solution formulations and is commonly used as a reference compound in hepatotoxicity and metabolism investigations.
  • Water-soluble prodrug of paracetamol, suitable for parenteral preparation.
  • Facilitates rapid conversion to active acetaminophen in vivo.
  • Available as solid and solution formulations for flexible dosing and sample preparation.
  • Used in studies of hepatotoxicity and cytochrome P450 metabolism.
  • Provided with datasheet and safety documentation for laboratory use.
Catalog No. 50-002-43259 Supplier Medchemexpress LLC Supplier No. HY108292100MG
Propacetamol (hydrochloride) is the hydrochloride salt of propacetamol, a water-soluble prodrug of paracetamol (acetaminophen) used as an analgesic in research settings. Supplied for biochemical and in vivo studies, it is provided in solid and solution formulations and is commonly used as a reference compound in hepatotoxicity and metabolism investigations.
  • Water-soluble prodrug of paracetamol, suitable for parenteral preparation.
  • Facilitates rapid conversion to active acetaminophen in vivo.
  • Available as solid and solution formulations for flexible dosing and sample preparation.
  • Used in studies of hepatotoxicity and cytochrome P450 metabolism.
  • Provided with datasheet and safety documentation for laboratory use.