Learn More
Medchemexpress LLC Propacetamol hydrochloride | 66532-86-3 | MFCD00941392 | >=98.0% | 300.79 | C14H21ClN2O3 | 250 MG
SDP
Catalog No. 5000242478
Encompass_Preferred
Propacetamol hydrochloride is the hydrochloride salt of propacetamol, a water-soluble prodrug of paracetamol (acetaminophen) supplied for research use. It is used as an analgesic and antipyretic reagent and in studies of prodrug metabolism, pharmacology, and analytical method development. This material is intended for laboratory research only and not for human or veterinary use.
  • Analgesic prodrug used to study paracetamol metabolism.
  • Suitable for antipyretic and pharmacology assays.
  • Available as solid and pre-made DMSO solutions.
  • Molecular formula C14H21ClN2O3; molecular weight 300.79.
  • Intended for research applications; not for clinical use.
Catalog No. 50-002-42478 Supplier Medchemexpress LLC Supplier No. HY108292250MG
Propacetamol hydrochloride is the hydrochloride salt of propacetamol, a water-soluble prodrug of paracetamol (acetaminophen) supplied for research use. It is used as an analgesic and antipyretic reagent and in studies of prodrug metabolism, pharmacology, and analytical method development. This material is intended for laboratory research only and not for human or veterinary use.
  • Analgesic prodrug used to study paracetamol metabolism.
  • Suitable for antipyretic and pharmacology assays.
  • Available as solid and pre-made DMSO solutions.
  • Molecular formula C14H21ClN2O3; molecular weight 300.79.
  • Intended for research applications; not for clinical use.