Learn More
Cayman Chemical Propacetamol (hydrochloride), Purity: ≥90%, CAS: 66532-86-3, 100 mg
SDP
Catalog No. 502911879
Encompass_Preferred
A prodrug form of acetaminophen; reduces acetic acid-induced writhing in mice and LPS-induced pyresis in rats at 200 and 600 mg/kg, respectively; induces hepatotoxicity, decreases hepatic GSH, SOD, and GPX levels, increases hepatic MDA and nitrotyrosine levels, and increases mortality in mice at 1200 mg/kg
Catalog No. 50-291-1879 Supplier Cayman Chemical Supplier No. 36134100 MG
A prodrug form of acetaminophen; reduces acetic acid-induced writhing in mice and LPS-induced pyresis in rats at 200 and 600 mg/kg, respectively; induces hepatotoxicity, decreases hepatic GSH, SOD, and GPX levels, increases hepatic MDA and nitrotyrosine levels, and increases mortality in mice at 1200 mg/kg