Learn More
Apexbio Technology LLC Propafenone HCl 34183-22-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090035
Encompass_Preferred
Propafenone hydrochloride (CAS 34183-22-7) is a synthetic small molecule classified as a Class Ic antiarrhythmic agent It acts by inhibiting fast inward sodium currents in cardiac myocytes resulting in slowed conduction and stabilization of cardiac electrical activity Through this sodium channel blockade Propafenone hydrochloride is utilized in biomedical research to investigate the electrophysiological properties of myocardial tissue and to model atrial and ventricular arrhythmias Its application aids in elucidating the molecular mechanisms underlying abnormal cardiac rhythms and supports the development of therapeutic interventions for tachyarrhythmic conditions
Catalog No. 50-000-90035 Supplier Apexbio Technology LLC Supplier No. B22815.1
Propafenone hydrochloride (CAS 34183-22-7) is a synthetic small molecule classified as a Class Ic antiarrhythmic agent It acts by inhibiting fast inward sodium currents in cardiac myocytes resulting in slowed conduction and stabilization of cardiac electrical activity Through this sodium channel blockade Propafenone hydrochloride is utilized in biomedical research to investigate the electrophysiological properties of myocardial tissue and to model atrial and ventricular arrhythmias Its application aids in elucidating the molecular mechanisms underlying abnormal cardiac rhythms and supports the development of therapeutic interventions for tachyarrhythmic conditions