Learn More
Apexbio Technology LLC Propafenone HCl 34183-22-7 1g
SDP
Catalog No. 5000090033
Encompass_Preferred
Propafenone hydrochloride (CAS 34183-22-7) is a synthetic small molecule classified as a Class Ic antiarrhythmic agent It acts by inhibiting fast inward sodium currents in cardiac myocytes resulting in slowed conduction and stabilization of cardiac electrical activity Through this sodium channel blockade Propafenone hydrochloride is utilized in biomedical research to investigate the electrophysiological properties of myocardial tissue and to model atrial and ventricular arrhythmias Its application aids in elucidating the molecular mechanisms underlying abnormal cardiac rhythms and supports the development of therapeutic interventions for tachyarrhythmic conditions
Catalog No. 50-000-90033 Supplier Apexbio Technology LLC Supplier No. B22811000
Add to Cart
Add to Cart
Propafenone hydrochloride (CAS 34183-22-7) is a synthetic small molecule classified as a Class Ic antiarrhythmic agent It acts by inhibiting fast inward sodium currents in cardiac myocytes resulting in slowed conduction and stabilization of cardiac electrical activity Through this sodium channel blockade Propafenone hydrochloride is utilized in biomedical research to investigate the electrophysiological properties of myocardial tissue and to model atrial and ventricular arrhythmias Its application aids in elucidating the molecular mechanisms underlying abnormal cardiac rhythms and supports the development of therapeutic interventions for tachyarrhythmic conditions