Learn More
Medchemexpress LLC Propafenone hydrochloride | 34183-22-7 | 98.5% | 1 G
SDP
Catalog No. 5000408600
Encompass_Preferred
Propafenone hydrochloride is a class of anti-arrhythmic medication used to treat illnesses associated with rapid heartbeats, such as atrial and ventricular arrhythmias. It works by slowing the influx of sodium ions into cardiac muscle cells, which decreases the excitability of these cells. This compound is more selective for cells with a high rate of activity but also affects normal cells more than Class Ia or Ib antiarrhythmics. It also possesses beta-adrenergic blocking activity, which can lead to bradycardia and bronchospasm.
  • Treats atrial and ventricular arrhythmias
  • Slows influx of sodium ions into cardiac muscle cells
  • Decreases excitability of cardiac cells
  • More selective for cells with a high rate of activity
  • Possesses beta-adrenergic blocking activity
Catalog No. 50-004-08600 Supplier Medchemexpress LLC Supplier No. HYB0432A1G
Propafenone hydrochloride is a class of anti-arrhythmic medication used to treat illnesses associated with rapid heartbeats, such as atrial and ventricular arrhythmias. It works by slowing the influx of sodium ions into cardiac muscle cells, which decreases the excitability of these cells. This compound is more selective for cells with a high rate of activity but also affects normal cells more than Class Ia or Ib antiarrhythmics. It also possesses beta-adrenergic blocking activity, which can lead to bradycardia and bronchospasm.
  • Treats atrial and ventricular arrhythmias
  • Slows influx of sodium ions into cardiac muscle cells
  • Decreases excitability of cardiac cells
  • More selective for cells with a high rate of activity
  • Possesses beta-adrenergic blocking activity