Learn More
Medchemexpress LLC Propafenone hydrochloride | 34183-22-7 | 98.5% | 5 G
SDP
Catalog No. 5000408598
Encompass_Preferred
Propafenone hydrochloride, also known as SA-79 hydrochloride, is an anti-arrhythmic medication used to treat illnesses involving rapid heartbeats such as atrial and ventricular arrhythmias. It functions by slowing the influx of sodium ions into cardiac muscle cells, thereby decreasing their excitability. Propafenone is more selective for cells with a high rate and blocks normal cells more effectively than Class Ia or Ib antiarrhythmics. Unlike typical Class Ic antiarrhythmics, it also exhibits beta-adrenergic blocker activity, which can lead to bradycardia and bronchospasm.
  • More selective for cells with a high rate
  • Blocks normal cells more effectively than Class Ia or Ib antiarrhythmics
  • Exhibits beta-adrenergic blocker activity
Catalog No. 50-004-08598 Supplier Medchemexpress LLC Supplier No. HYB0432A5G
Propafenone hydrochloride, also known as SA-79 hydrochloride, is an anti-arrhythmic medication used to treat illnesses involving rapid heartbeats such as atrial and ventricular arrhythmias. It functions by slowing the influx of sodium ions into cardiac muscle cells, thereby decreasing their excitability. Propafenone is more selective for cells with a high rate and blocks normal cells more effectively than Class Ia or Ib antiarrhythmics. Unlike typical Class Ic antiarrhythmics, it also exhibits beta-adrenergic blocker activity, which can lead to bradycardia and bronchospasm.
  • More selective for cells with a high rate
  • Blocks normal cells more effectively than Class Ia or Ib antiarrhythmics
  • Exhibits beta-adrenergic blocker activity