Learn More
Cayman Chemical Propafenone (hydrochloride), Purity: ≥98%, CAS: 34183-22-7, 10 g
SDP
Catalog No. 502985990
Encompass_Preferred
A class I antiarrhythmic agent; inhibits ICa in isolated guinea pig ventricular myocytes (IC50 = 5 µM), as well as IKATP in isolated rabbit atrial and ventricular monocytes (IC50s = 1.26 and 4.94 µM, respectively); blocks INa, IK, and transient outward potassium currents in various cardiac cells; reverses sinus rhythm in a dog model of barium chloride-induced ventricular arrhythmia at 2 and 3 mg/kg, i.v.
Catalog No. 50-298-5990 Supplier Cayman Chemical Supplier No. 2187110 G
A class I antiarrhythmic agent; inhibits ICa in isolated guinea pig ventricular myocytes (IC50 = 5 µM), as well as IKATP in isolated rabbit atrial and ventricular monocytes (IC50s = 1.26 and 4.94 µM, respectively); blocks INa, IK, and transient outward potassium currents in various cardiac cells; reverses sinus rhythm in a dog model of barium chloride-induced ventricular arrhythmia at 2 and 3 mg/kg, i.v.