Learn More
R&D Systems™ ProPak Recombinant Human IL-15 GMP Protein
Description
Specifications
Specifications
| Accession Number | P40933.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS, recombinant HSA, and Trehalose. |
| Gene ID (Entrez) | 3600 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 9 kDa, under reducing conditions., MolecularWeight-theroretical: 13 kDa |
| Quantity | 10 μg |
| Source | E. coli-derived human IL-15 protein Asn49-Ser162 Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 6 months when stored between -14 °C and -40 °C. Can be stored up to 2 weeks at 2-8 °C. |
| Endotoxin Concentration | <5.0 EU/mL by the LAL method. |
| Gene Alias | IL15, IL-15MGC9721, interleukin 15, interleukin-15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.