Learn More
R&D Systems™ ProPak™ Recombinant Human IL-2 GMP Protein, CF
Description
Specifications
Specifications
| Accession Number | P60568.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS, recombinant HSA, and Trehalose. |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | M.W.-observed: 13 kDa, under reducing conditions., M.W.-theroretical: 15.5 kDa |
| Quantity | 150,000 IU |
| Source | E. coli-derived human IL-2 protein Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidlelines. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 6 months when stored between -14 °C and -40 °C. Can be stored up to 2 weeks at 2-8 °C. Refer to lot specific COA for the Use by Date. |
| Endotoxin Concentration | <5.0 EU/mL by the LAL method. |
| Gene Alias | aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.