Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propamidine isethionate | 140-63-6 | MFCD01728620 | 95.7% | 564.63 g/mol | C21H32N4O10S2 | 5 MG
SDP

Catalog No. 5000262374
Encompass_Preferred

Propamidine isethionate is a diamidine-class research reagent supplied as the isethionate salt and used primarily in laboratory studies such as Acanthamoeba keratitis research. It is provided as a high-purity solid intended for experimental use.

  • Diamidine-class compound used in antiparasitic research.
  • Applicable to Acanthamoeba keratitis research.
  • Purity approximately 95.7%.
  • Molecular weight 564.63 g/mol.
  • Chemical formula C21H32N4O10S2.
  • White to off-white solid appearance.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small milligram quantities for laboratory use.

Catalog No. 50-002-62374 Supplier Medchemexpress LLC Supplier No. HY130402A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Propamidine isethionate is a diamidine-class research reagent supplied as the isethionate salt and used primarily in laboratory studies such as Acanthamoeba keratitis research. It is provided as a high-purity solid intended for experimental use.

  • Diamidine-class compound used in antiparasitic research.
  • Applicable to Acanthamoeba keratitis research.
  • Purity approximately 95.7%.
  • Molecular weight 564.63 g/mol.
  • Chemical formula C21H32N4O10S2.
  • White to off-white solid appearance.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small milligram quantities for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.