Learn More
Cayman Chemical Propamocarb, Purity: ≥98%, CAS: 24579-73-5, 100 mg
SDP
Catalog No. 502918933
Encompass_Preferred
A carbamate fungicide; active against the plant pathogenic fungi P. aphanidermatum, P. dissotocum, P. irregulare, and P. ultimum at 400 µg/ml; potentiates estrogen receptor-mediated transactivation induced by 17β-estradiol in MCF-7 cells in a reporter assay but has no effect when used alone; has been found in spring water
Catalog No. 50-291-8933 Supplier Cayman Chemical Supplier No. 39502100 MG
A carbamate fungicide; active against the plant pathogenic fungi P. aphanidermatum, P. dissotocum, P. irregulare, and P. ultimum at 400 µg/ml; potentiates estrogen receptor-mediated transactivation induced by 17β-estradiol in MCF-7 cells in a reporter assay but has no effect when used alone; has been found in spring water