Learn More
Medchemexpress LLC Propamocarb-d7 | 1398065-89-8 | 1 MG
SDP
Catalog No. 5000411238
Encompass_Preferred
Propamocarb-d7 is the deuterium-labeled version of Propamocarb, a systemic fungicide widely used to protect plants such as cucumbers and tomatoes from pathogens. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11238 Supplier Medchemexpress LLC Supplier No. HYB2026S1MG
Propamocarb-d7 is the deuterium-labeled version of Propamocarb, a systemic fungicide widely used to protect plants such as cucumbers and tomatoes from pathogens. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.