Learn More
Medchemexpress LLC Propamocarb-d7 | 1398065-89-8 | 5 MG
SDP
Catalog No. 5000411229
Encompass_Preferred
Propamocarb-d7 is the deuterium-labeled version of Propamocarb, a systemic fungicide used to protect plants such as cucumbers and tomatoes from pathogens.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11229 Supplier Medchemexpress LLC Supplier No. HYB2026S5MG
Propamocarb-d7 is the deuterium-labeled version of Propamocarb, a systemic fungicide used to protect plants such as cucumbers and tomatoes from pathogens.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.