Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propan-2-one oxime | 127-06-0 | 99.9% | 73.09 | 1 KG
SDP

Catalog No. 5000451283
Encompass_Preferred

Propan-2-one oxime is a biological material or organic compound widely used in life science research.

  • For research use only.
  • Appearance: Solid
  • Color: White to off-white
  • Store powder at -20°C for 3 years, 4°C for 2 years.
  • Store in solvent at -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-51283 Supplier Medchemexpress LLC Supplier No. HYW0088401KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propan-2-one oxime is a biological material or organic compound widely used in life science research.

  • For research use only.
  • Appearance: Solid
  • Color: White to off-white
  • Store powder at -20°C for 3 years, 4°C for 2 years.
  • Store in solvent at -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.