Learn More
Medchemexpress LLC Propan-2-one oxime | 127-06-0 | 99.97% | 73.09 | 50 G
SDP
Catalog No. 5000453725
Encompass_Preferred
Propan-2-one oxime is a biological material or organic compound widely used in life science research. It is presented as a white to off-white solid and is intended for research use only, not for patient use.
  • Biological material or organic compound
  • Wide application in life science research
  • White to off-white solid appearance
  • For research use only
  • Store as powder at -20°C for up to 3 years or at 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature in continental US
  • Technical documentation available (data sheet, COA, SDS, HNMR, GC, and handling instructions)
Catalog No. 50-004-53725 Supplier Medchemexpress LLC Supplier No. HYW00884050G
Propan-2-one oxime is a biological material or organic compound widely used in life science research. It is presented as a white to off-white solid and is intended for research use only, not for patient use.
  • Biological material or organic compound
  • Wide application in life science research
  • White to off-white solid appearance
  • For research use only
  • Store as powder at -20°C for up to 3 years or at 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature in continental US
  • Technical documentation available (data sheet, COA, SDS, HNMR, GC, and handling instructions)