Learn More
Medchemexpress LLC Propan-2-yl 2-sulfanylacetate | 7383-61-1 | MFCD00022078 | 134.20 g/mol | C5H10O2S | 100 MG
SDP
Catalog No. 5000283196
Encompass_Preferred
Propan-2-yl 2-sulfanylacetate (isopropyl thioglycolate) is an organosulfur compound supplied for chemical research and industrial formulations. It is commonly used as a stabilizer for dyes, an additive in polymer flame-retardant systems, and as a component in metal-cleaning agents where thioglycolate functionality is required.
  • Organosulfur reagent useful as a dye stabilizer.
  • Additive for polymer flame-retardant formulations.
  • Component for metal cleaning and surface treatment agents.
  • Molecular formula C5H10O2S; molecular weight 134.20 g/mol; CAS 7383-61-1.
  • Available in small lab-scale quantities for research use.
Catalog No. 50-002-83196 Supplier Medchemexpress LLC Supplier No. HYW200537100MG
Add to Cart
Add to Cart
Propan-2-yl 2-sulfanylacetate (isopropyl thioglycolate) is an organosulfur compound supplied for chemical research and industrial formulations. It is commonly used as a stabilizer for dyes, an additive in polymer flame-retardant systems, and as a component in metal-cleaning agents where thioglycolate functionality is required.
  • Organosulfur reagent useful as a dye stabilizer.
  • Additive for polymer flame-retardant formulations.
  • Component for metal cleaning and surface treatment agents.
  • Molecular formula C5H10O2S; molecular weight 134.20 g/mol; CAS 7383-61-1.
  • Available in small lab-scale quantities for research use.