Learn More
Medchemexpress LLC Propanal, 2-methyl-2-(methylsulfonyl)-, O-[(methylamino)carbonyl]oxime | 1646-88-4 | 99.5% | 222.26 | 25 MG
SDP
Catalog No. 5000384614
Encompass_Preferred
Aldicarb sulfone is a metabolite of the carbamate pesticide Aldicarb, exhibiting insecticidal activity. It also acts as a cholinesterase inhibitor and is for research use only.
  • Inhibits egg hatching and reduces nematode motility.
  • Hinders male larval development in Tylenchulus semipenetrans.
  • Decreases plasma pseudocholinesterase levels.
  • Decreases red blood cell cholinesterase levels.
Catalog No. 50-003-84614 Supplier Medchemexpress LLC Supplier No. 25MGHY17530
Aldicarb sulfone is a metabolite of the carbamate pesticide Aldicarb, exhibiting insecticidal activity. It also acts as a cholinesterase inhibitor and is for research use only.
  • Inhibits egg hatching and reduces nematode motility.
  • Hinders male larval development in Tylenchulus semipenetrans.
  • Decreases plasma pseudocholinesterase levels.
  • Decreases red blood cell cholinesterase levels.