Learn More
Medchemexpress LLC Propanal, 3-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]- | 1807530-10-4 | 95.0% | 231.25 g/mol | C9H17N3O4 | 5 MG
SDP
Catalog No. 5000222858
Encompass_Preferred
Azido-PEG3-aldehyde is an azide-functionalized PEG3 aldehyde linker used in chemical synthesis and bioconjugation. It combines an azide handle for click chemistry with an aldehyde for further conjugation, providing a short PEG spacer to improve solubility and flexibility in bifunctional molecules. Supplied as a high-purity research reagent for laboratory use.
  • Contains azide and aldehyde functional groups for orthogonal conjugation.
  • Provides a three-unit polyethylene glycol spacer to increase solubility and flexibility.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and reductive amination.
  • High purity suitable for research use (≥95.0%).
  • Stable when stored cold; follow recommended storage for long-term stability.
Catalog No. 50-002-22858 Supplier Medchemexpress LLC Supplier No. HY1385185MG
Azido-PEG3-aldehyde is an azide-functionalized PEG3 aldehyde linker used in chemical synthesis and bioconjugation. It combines an azide handle for click chemistry with an aldehyde for further conjugation, providing a short PEG spacer to improve solubility and flexibility in bifunctional molecules. Supplied as a high-purity research reagent for laboratory use.
  • Contains azide and aldehyde functional groups for orthogonal conjugation.
  • Provides a three-unit polyethylene glycol spacer to increase solubility and flexibility.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and reductive amination.
  • High purity suitable for research use (≥95.0%).
  • Stable when stored cold; follow recommended storage for long-term stability.