Learn More
Medchemexpress LLC Propanamide, 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]- | 52806-53-8 | 99.8% | 292.21 | 100 MG
SDP
Catalog No. 5000454240
Encompass_Preferred
Hydroxyflutamide (HFT) is an orally active metabolite of Flutamide, functioning as a potent androgen receptor antagonist with an IC50 of 700 nM. This compound has demonstrated effects on embryonic and reproductive tract development in mice and can enhance the efficacy of Bacillus Calmette-Guérin (BCG) in inhibiting bladder cancer progression. It is utilized in research concerning tumors and reproductive diseases, impacting targets such as Akt, PI3K, Gap Junction Protein, and Integrin.
  • Potent androgen receptor antagonist with an IC50 of 700 nM
  • Orally active metabolite
  • Influences embryonic and reproductive tract development
  • Enhances Bacillus Calmette-Guérin (BCG) efficacy against bladder cancer
  • Useful in tumor and reproductive disease research
Catalog No. 50-004-54240 Supplier Medchemexpress LLC Supplier No. HYW013272100MG
Hydroxyflutamide (HFT) is an orally active metabolite of Flutamide, functioning as a potent androgen receptor antagonist with an IC50 of 700 nM. This compound has demonstrated effects on embryonic and reproductive tract development in mice and can enhance the efficacy of Bacillus Calmette-Guérin (BCG) in inhibiting bladder cancer progression. It is utilized in research concerning tumors and reproductive diseases, impacting targets such as Akt, PI3K, Gap Junction Protein, and Integrin.
  • Potent androgen receptor antagonist with an IC50 of 700 nM
  • Orally active metabolite
  • Influences embryonic and reproductive tract development
  • Enhances Bacillus Calmette-Guérin (BCG) efficacy against bladder cancer
  • Useful in tumor and reproductive disease research