Learn More
Medchemexpress LLC Propanamide, 3-[[3-[4-[[3-(β-D-glucopyranosyloxy)-5-(1-methylethyl)-1H-pyrazol-4-yl]methyl]-3-methylphenox... | 666842-36-0 | 98.8% | 536.62 g·mol⁻¹ | C26H40N4O8 | 5MG
SDP
Catalog No. 5000235442
Encompass_Preferred
KGA-2727 is a selective, high-affinity, orally active inhibitor of the sodium-glucose cotransporter 1 (SGLT1) used in research on glucose transport and metabolic effects. It exhibits Ki values in the low nanomolar range for human and rat SGLT1 and has demonstrated antidiabetic efficacy in preclinical studies.
  • Selective SGLT1 inhibition with low nanomolar Ki values.
  • High affinity and oral activity observed in preclinical models.
  • Demonstrated antidiabetic efficacy in rodent studies.
  • Supplied at high purity (≈98.8%), suitable for research use.
  • Offered as solid packs and ready-to-use DMSO solution formats.
  • Storage guidance provided for solid and solution stability.
Catalog No. 50-002-35442 Supplier Medchemexpress LLC Supplier No. HY1237975MG
KGA-2727 is a selective, high-affinity, orally active inhibitor of the sodium-glucose cotransporter 1 (SGLT1) used in research on glucose transport and metabolic effects. It exhibits Ki values in the low nanomolar range for human and rat SGLT1 and has demonstrated antidiabetic efficacy in preclinical studies.
  • Selective SGLT1 inhibition with low nanomolar Ki values.
  • High affinity and oral activity observed in preclinical models.
  • Demonstrated antidiabetic efficacy in rodent studies.
  • Supplied at high purity (≈98.8%), suitable for research use.
  • Offered as solid packs and ready-to-use DMSO solution formats.
  • Storage guidance provided for solid and solution stability.