Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanamide, 3-[3,5-bis(trifluoromethyl)phenoxy]-N-[4-cyano-3-(trifluoromethyl)phenyl] | 1928724-23-5 | MFCD32174353 | 99.98% | 500.31 g/mol | C20H13F9N2O3 | 5 MG
SDP

Catalog No. 5000222337
Encompass_Preferred

SK33 is a trifluoromethylated enobosarm analog that acts as a potent, tissue-selective anti-androgen by reducing androgen receptor (AR) transcriptional activity. It is intended for research use in studies of AR signaling and nuclear receptor pathways.

  • Potent, tissue-selective anti-androgen activity.
  • Reduces androgen receptor (AR) transcriptional activity.
  • Target: androgen receptor (AR).
  • Pathway: vitamin D related / nuclear receptor.
  • Molecular formula: C20H13F9N2O3.
  • Molecular weight: 500.31 g/mol.
  • Purity: 99.98% (LCMS).
  • CAS number: 1928724-23-5.
  • Available in a 5 mg quantity for laboratory research.

Catalog No. 50-002-22337 Supplier Medchemexpress LLC Supplier No. HY1357325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SK33 is a trifluoromethylated enobosarm analog that acts as a potent, tissue-selective anti-androgen by reducing androgen receptor (AR) transcriptional activity. It is intended for research use in studies of AR signaling and nuclear receptor pathways.

  • Potent, tissue-selective anti-androgen activity.
  • Reduces androgen receptor (AR) transcriptional activity.
  • Target: androgen receptor (AR).
  • Pathway: vitamin D related / nuclear receptor.
  • Molecular formula: C20H13F9N2O3.
  • Molecular weight: 500.31 g/mol.
  • Purity: 99.98% (LCMS).
  • CAS number: 1928724-23-5.
  • Available in a 5 mg quantity for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.