Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanamide, N-[cis-4-[2-[[4-[4-[6-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1-oxo-1H-isoindol-5-yl] | 2953426-43-0 | 98.0% | 855.98 | C47H53N9O7 | 5 MG
SDP

Catalog No. 5000222303
Encompass_Preferred

A threonine tyrosine kinase (TTK)-targeting PROTAC degrader developed for research use, showing potent target degradation in cancer cell lines and antitumor activity in xenograft models. Supplied as a light yellow solid with documented analytical and storage data.

  • Potent TTK degradation with reported DC50 values of 1.7 nM (COLO-205) and 5.8 nM (HCT-116).
  • High reported purity of 98.01%.
  • Molecular weight 855.98 and formula C47H53N9O7.
  • Available in small-molecule quantities, including 5 mg solid format.
  • Recommended storage: 4°C protected from light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-22303 Supplier Medchemexpress LLC Supplier No. HY1439045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A threonine tyrosine kinase (TTK)-targeting PROTAC degrader developed for research use, showing potent target degradation in cancer cell lines and antitumor activity in xenograft models. Supplied as a light yellow solid with documented analytical and storage data.

  • Potent TTK degradation with reported DC50 values of 1.7 nM (COLO-205) and 5.8 nM (HCT-116).
  • High reported purity of 98.01%.
  • Molecular weight 855.98 and formula C47H53N9O7.
  • Available in small-molecule quantities, including 5 mg solid format.
  • Recommended storage: 4°C protected from light; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.