Learn More
Medchemexpress LLC Propane-1,3-diol | 504-63-2 | MFCD00002949 | 98.8% | 76.09 g/mol | C3H8O2 | 500 G
SDP
Catalog No. 5000300646
Encompass_Preferred
1,3-Propanediol (propane-1,3-diol; CAS 504-63-2) is a small, water-miscible diol supplied for research and analytical use. It is produced naturally via glycerol fermentation and is used as a solvent, building block in polymer synthesis, and a reagent in biological studies. For research use only.
Catalog No. 50-003-00646 Supplier Medchemexpress LLC Supplier No. HYW017758500G
1,3-Propanediol (propane-1,3-diol; CAS 504-63-2) is a small, water-miscible diol supplied for research and analytical use. It is produced naturally via glycerol fermentation and is used as a solvent, building block in polymer synthesis, and a reagent in biological studies. For research use only.