Learn More
Cambridge Isotope Laboratories PROPANE (D8, 98%), 1 L, 2875-94-7
Supplier:  Cambridge Isotope Laboratories DLM1531LB
Encompass
PROPANE (D8, 98%), 1 L, 2875-94-* This item is shipped in a Code K-460 stainless steel cylinder. The fee for the cylinder will need added to the order as a separate charge, per each. Please include part# NC1924736to the order for the cylinder fee.
Catalog No. 50-163-0887