Learn More
Medchemexpress LLC Propane sultone | 1120-71-4 | 100.0% | 122.14 | 25 G
SDP
Catalog No. 5000367101
Encompass_Preferred
Propane sultone is a multifunctional intermediate used in the synthesis of other active compounds and as an additive in lithium battery electrolytes.
  • Can be combined with cellulose and carbohydrate.
  • Used for sizing, printing, and textile applications.
  • Used for the manufacture of dyes.
Catalog No. 50-003-67101 Supplier Medchemexpress LLC Supplier No. 25GHYW010479
Propane sultone is a multifunctional intermediate used in the synthesis of other active compounds and as an additive in lithium battery electrolytes.
  • Can be combined with cellulose and carbohydrate.
  • Used for sizing, printing, and textile applications.
  • Used for the manufacture of dyes.