Learn More
Medchemexpress LLC Propane sultone | 1120-71-4 | 99.9% | 122.14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW010479100G
Encompass_Preferred
Propane sultone is a versatile intermediate that can be employed in the synthesis of various active compounds. It also serves as an additive in lithium battery electrolytes.
  • Multifunctional intermediate
  • Can be used for the synthesis of other active compounds
  • Can be used as an additive in lithium battery electrolytes
  • Combines with cellulose and carbohydrate for use in sizing, printing, and textiles
  • Can be used for the manufacture of dyes
Catalog No. 50-003-66458