Learn More
Medchemexpress LLC Propane sultone | 1120-71-4 | 99.9% | 122.14 | 500 G
SDP
Catalog No. 5000366451
Encompass_Preferred
Propane sultone is a multifunctional intermediate used for synthesizing other active compounds and as an additive in lithium battery electrolytes. It is also used in various industrial applications such as sizing, printing, textiles, and the manufacture of dyes.
  • Used for the synthesis of other active compounds
  • Can be an additive in lithium battery electrolytes
  • Combines with cellulose and carbohydrate
  • Suitable for sizing, printing, and textile applications
  • Used in the manufacture of dyes
Catalog No. 50-003-66451 Supplier Medchemexpress LLC Supplier No. HYW010479500G
Propane sultone is a multifunctional intermediate used for synthesizing other active compounds and as an additive in lithium battery electrolytes. It is also used in various industrial applications such as sizing, printing, textiles, and the manufacture of dyes.
  • Used for the synthesis of other active compounds
  • Can be an additive in lithium battery electrolytes
  • Combines with cellulose and carbohydrate
  • Suitable for sizing, printing, and textile applications
  • Used in the manufacture of dyes