Learn More
Medchemexpress LLC Propanedinitrile, 2-[(3,4-dihydroxy-5-nitrophenyl)methylene]- | 116313-73-6 | 98.7% | 231.16 g·mol⁻¹ | C10H5N3O4 | 50 MG
SDP
Catalog No. 5000245870
Encompass_Preferred
Tyrphostin AG 1288 is a research-grade small-molecule tyrosine kinase inhibitor reported to inhibit ICAM1 expression and TNFα-mediated cytotoxicity in vitro. The compound is supplied with analytical characterization and recommended storage conditions to support reliable laboratory use.
  • Potent tyrosine kinase inhibitor in biochemical and cellular assays.
  • Inhibits ICAM1 expression in cell-based studies.
  • Reduces TNFα-mediated cytotoxicity in vitro.
  • High purity suitable for research applications (≈98.7%).
  • Available in multiple small pack sizes for laboratory workflows.
  • Stable when stored under recommended conditions for powder and solutions.
Catalog No. 50-002-45870 Supplier Medchemexpress LLC Supplier No. HY10196150MG
Tyrphostin AG 1288 is a research-grade small-molecule tyrosine kinase inhibitor reported to inhibit ICAM1 expression and TNFα-mediated cytotoxicity in vitro. The compound is supplied with analytical characterization and recommended storage conditions to support reliable laboratory use.
  • Potent tyrosine kinase inhibitor in biochemical and cellular assays.
  • Inhibits ICAM1 expression in cell-based studies.
  • Reduces TNFα-mediated cytotoxicity in vitro.
  • High purity suitable for research applications (≈98.7%).
  • Available in multiple small pack sizes for laboratory workflows.
  • Stable when stored under recommended conditions for powder and solutions.