Learn More
Medchemexpress LLC Propanedinitrile, 2-[(3,4-dihydroxy-5-nitrophenyl)methylene] | 116313-73-6 | MFCD00236451 | 98.7% | 231.16 g/mol | C10H5N3O4 | 5 MG
SDP
Catalog No. 5000248008
Encompass_Preferred
Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in research to modulate signaling pathways. Supplied as a light yellow solid, it inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro and is characterized by formula C10H5N3O4, molecular weight 231.16 g/mol, and typical purity of approximately 98.7%.
  • Potent tyrosine kinase inhibition in cellular assays.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity.
  • High purity suitable for biochemical and cellular studies.
  • Available in small milligram-scale packages for laboratory use.
  • Solid form with defined appearance for straightforward handling.
Catalog No. 50-002-48008 Supplier Medchemexpress LLC Supplier No. HY1019615MG
Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in research to modulate signaling pathways. Supplied as a light yellow solid, it inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro and is characterized by formula C10H5N3O4, molecular weight 231.16 g/mol, and typical purity of approximately 98.7%.
  • Potent tyrosine kinase inhibition in cellular assays.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity.
  • High purity suitable for biochemical and cellular studies.
  • Available in small milligram-scale packages for laboratory use.
  • Solid form with defined appearance for straightforward handling.