Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanedioic acid, 2-oxo-, 1,3-diethyl ester | 609-09-6 | MFCD00009121 | 99.5% | 174.15 g/mol | C7H10O5 | 10 G
SDP

Catalog No. 5000301226
Encompass_Preferred

1,3-Diethyl 2-oxopropanedioate is a ketomalonate ester used as an organic reagent and biochemical building block in research. It is a colorless to light yellow liquid with high purity, suitable for organic synthesis and analytical applications.

  • High purity (99.5% by GC) suitable for synthetic use.
  • Colorless to light yellow liquid physical form.
  • Molecular formula C7H10O5.
  • Molecular weight 174.15 g/mol.
  • Serves as a versatile building block in organic synthesis.
  • Used in enolate and acylation chemistry and heterocycle formation.
  • Supplied in small research pack sizes for laboratory use.

Catalog No. 50-003-01226 Supplier Medchemexpress LLC Supplier No. HYW01680210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Diethyl 2-oxopropanedioate is a ketomalonate ester used as an organic reagent and biochemical building block in research. It is a colorless to light yellow liquid with high purity, suitable for organic synthesis and analytical applications.

  • High purity (99.5% by GC) suitable for synthetic use.
  • Colorless to light yellow liquid physical form.
  • Molecular formula C7H10O5.
  • Molecular weight 174.15 g/mol.
  • Serves as a versatile building block in organic synthesis.
  • Used in enolate and acylation chemistry and heterocycle formation.
  • Supplied in small research pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.