Learn More
Medchemexpress LLC Propanedioic acid, sodium salt, hydrate (1:2:1) | 26522-85-0 | MFCD00002708 | 99.1% | 166.04 | C3H4Na2O5 | 25 G
SDP
Catalog No. 5000304104
Encompass_Preferred
Sodium malonate hydrate is the disodium salt of malonic acid (monohydrate) used as a reagent and complexing agent in analytical and life-science applications. It is commonly applied for determination of beryllium and copper, and in biochemical syntheses where a high-purity malonate source is required. The material is supplied as a white to off-white solid with documented assay and moisture specifications.
Catalog No. 50-003-04104 Supplier Medchemexpress LLC Supplier No. 25GHYW017165
Sodium malonate hydrate is the disodium salt of malonic acid (monohydrate) used as a reagent and complexing agent in analytical and life-science applications. It is commonly applied for determination of beryllium and copper, and in biochemical syntheses where a high-purity malonate source is required. The material is supplied as a white to off-white solid with documented assay and moisture specifications.