Learn More
Medchemexpress LLC Propanenitrile, 3-(methylsulfonyl)- | 54863-37-5 | 99.9% | 133.17 | 250 MG
SDP
Catalog No. 5000385455
Encompass_Preferred
Dapansutrile is a potent, orally active, and selective NLRP3 inflammasome inhibitor. It exhibits anti-inflammatory activity and reduces immune factor levels. This product is intended for research use in inflammatory diseases.
  • Potent and selective NLRP3 inflammasome inhibitor
  • Orally active
  • Exhibits anti-inflammatory activity
  • Decreases immune factor levels
  • Used for research on inflammatory diseases
Catalog No. 50-003-85455 Supplier Medchemexpress LLC Supplier No. 250MGHY17629
Dapansutrile is a potent, orally active, and selective NLRP3 inflammasome inhibitor. It exhibits anti-inflammatory activity and reduces immune factor levels. This product is intended for research use in inflammatory diseases.
  • Potent and selective NLRP3 inflammasome inhibitor
  • Orally active
  • Exhibits anti-inflammatory activity
  • Decreases immune factor levels
  • Used for research on inflammatory diseases