Learn More
Medchemexpress LLC Propanenitrile, 3-(methylsulfonyl)- | 54863-37-5 | MFCD28557579 | 99.9% | 133.17 | 100 MG
SDP
Catalog No. 5000387482
Encompass_Preferred
Dapansutrile is a potent, orally active, and selective NLRP3 inflammasome inhibitor. It exhibits anti-inflammatory activity and reduces immune factor levels, making it suitable for research into inflammatory diseases.
  • Potent, orally active, and selective NLRP3 inflammasome inhibitor.
  • Exhibits anti-inflammatory activity.
  • Decreases immune factor levels.
  • Used for research of inflammatory diseases.
Catalog No. 50-003-87482 Supplier Medchemexpress LLC Supplier No. HY17629100MG
Dapansutrile is a potent, orally active, and selective NLRP3 inflammasome inhibitor. It exhibits anti-inflammatory activity and reduces immune factor levels, making it suitable for research into inflammatory diseases.
  • Potent, orally active, and selective NLRP3 inflammasome inhibitor.
  • Exhibits anti-inflammatory activity.
  • Decreases immune factor levels.
  • Used for research of inflammatory diseases.