Learn More
Medchemexpress LLC Propanidid 10mg | 1421-14-3 | 337.41 g/mol | C18H27NO5 | 10 MG
SDP
Catalog No. 5000227219
Encompass_Preferred
Propanidid is a short-acting, non-barbiturate general anesthetic and γ-aminobutyric acid type A (GABAA) receptor agonist supplied for research use. This 10 mg package is intended for laboratory studies; the compound has molecular formula C18H27NO5, molecular weight 337.41 g/mol, and reported purity of 98.19%.
  • Short-acting GABAA receptor agonist suitable for anesthetic pharmacology research
  • Provided as a 10 mg research-grade solid for small-scale experiments
  • High listed purity (98.19%) supports reproducible results
  • Known molecular formula C18H27NO5 and molecular weight 337.41 g/mol
  • Requires appropriate handling and storage per safety protocols
Catalog No. 50-002-27219 Supplier Medchemexpress LLC Supplier No. HY11645710MG
Propanidid is a short-acting, non-barbiturate general anesthetic and γ-aminobutyric acid type A (GABAA) receptor agonist supplied for research use. This 10 mg package is intended for laboratory studies; the compound has molecular formula C18H27NO5, molecular weight 337.41 g/mol, and reported purity of 98.19%.
  • Short-acting GABAA receptor agonist suitable for anesthetic pharmacology research
  • Provided as a 10 mg research-grade solid for small-scale experiments
  • High listed purity (98.19%) supports reproducible results
  • Known molecular formula C18H27NO5 and molecular weight 337.41 g/mol
  • Requires appropriate handling and storage per safety protocols