Learn More
Medchemexpress LLC Propanoic-13C sodium | 62601-06-3 | MFCD00002758 | 99.6% | 97.05 g/mol | 13C C3H5NaO2 | 10 MG
SDP
Catalog No. 5000297242
Encompass_Preferred
Sodium propionate labeled with 13C is the isotopically labeled sodium salt of propionic (propanoic) acid supplied for use as a tracer or internal standard in analytical chemistry (NMR, GC-MS, LC-MS). It is provided as a white solid for research use only and should be stored sealed, away from moisture and light.
Catalog No. 50-002-97242 Supplier Medchemexpress LLC Supplier No. HYB1773AS10MG
Sodium propionate labeled with 13C is the isotopically labeled sodium salt of propionic (propanoic) acid supplied for use as a tracer or internal standard in analytical chemistry (NMR, GC-MS, LC-MS). It is provided as a white solid for research use only and should be stored sealed, away from moisture and light.