Learn More
Medchemexpress LLC Propanoic-3-13C acid, 2-hydroxy-, monosodium salt, (2S)- | 201595-70-2 | 98.7% | 113.05 | C2(13C)H5NaO3 | 10 MG
SDP
Catalog No. 5000296808
Encompass_Preferred
13C-labelled L-lactic acid sodium supplied as a 20% w/w aqueous solution for use in metabolic tracing and analytical applications. The product is provided as a 10 mg aliquot of the labeled sodium salt, with reported purity of 98.7% and molecular weight 113.05. Store frozen at -20°C and protect from light; consult the manufacturer's datasheet/COA for handling and safety information.
  • Single 13C label at the 1-position for tracer studies.
  • Provided as a 20% w/w aqueous solution for convenient use.
  • High chemical purity suitable for analytical applications.
  • Ready-to-use sodium salt form for compatibility with biological buffers.
  • Store at -20°C and protect from light to maintain stability.
Catalog No. 50-002-96808 Supplier Medchemexpress LLC Supplier No. HYB2227BS310MG
13C-labelled L-lactic acid sodium supplied as a 20% w/w aqueous solution for use in metabolic tracing and analytical applications. The product is provided as a 10 mg aliquot of the labeled sodium salt, with reported purity of 98.7% and molecular weight 113.05. Store frozen at -20°C and protect from light; consult the manufacturer's datasheet/COA for handling and safety information.
  • Single 13C label at the 1-position for tracer studies.
  • Provided as a 20% w/w aqueous solution for convenient use.
  • High chemical purity suitable for analytical applications.
  • Ready-to-use sodium salt form for compatibility with biological buffers.
  • Store at -20°C and protect from light to maintain stability.