Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 2,3-dihydroxy-, monosodium salt, (R)- | 109175-64-6 | 99.0% | 128.06 g/mol | C3H5NaO4 | 5 MG
SDP

Catalog No. 5000223012
Encompass_Preferred

D-Glyceric acid sodium is the monosodium salt of D-glyceric acid, an endogenous metabolite used as an analytical standard and research reagent in metabolic and biochemical studies. It is supplied as a solid sodium salt suitable for assay development, metabolite profiling, and related biochemical experiments.

  • Sodium salt of D-glyceric acid, water-soluble solid.
  • CAS number 109175-64-6 for precise identification.
  • Molecular formula C3H5NaO4 and molecular weight 128.06 g/mol.
  • High purity (99.0%) suitable for analytical use.
  • Available in small research pack sizes, including 5 mg.
  • Commonly used in metabolic disorder and urine metabolite studies.

Catalog No. 50-002-23012 Supplier Medchemexpress LLC Supplier No. HY139070A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glyceric acid sodium is the monosodium salt of D-glyceric acid, an endogenous metabolite used as an analytical standard and research reagent in metabolic and biochemical studies. It is supplied as a solid sodium salt suitable for assay development, metabolite profiling, and related biochemical experiments.

  • Sodium salt of D-glyceric acid, water-soluble solid.
  • CAS number 109175-64-6 for precise identification.
  • Molecular formula C3H5NaO4 and molecular weight 128.06 g/mol.
  • High purity (99.0%) suitable for analytical use.
  • Available in small research pack sizes, including 5 mg.
  • Commonly used in metabolic disorder and urine metabolite studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.