Learn More
Medchemexpress LLC Propanoic acid, 2-(4-methoxyphenoxy)- | 13794-15-5 | MFCD01310545 | 99.8% | 196.20 g·mol⁻¹ | C10H12O4 | 250 MG
SDP
Catalog No. 5000303500
Encompass_Preferred
Lactisole free acid is a small-molecule antagonist of the sweet taste receptor T1R3 used to inhibit sweet taste signaling and study glucose-sensing pathways in cellular and pharmacological research. Supplied for research use only in solid and solution formats, it is commonly used in taste receptor assays and related in vitro studies.
  • Antagonist of the T1R3 sweet taste receptor.
  • Used for in vitro taste receptor and cellular signaling assays.
  • Available as solid (250 mg to 1 g) and 10 mM DMSO solution formats.
  • High purity: 99.8%.
  • Molecular formula C10H12O4, molecular weight 196.20 g·mol⁻¹.
  • For research use only; not for human consumption.
Catalog No. 50-003-03500 Supplier Medchemexpress LLC Supplier No. 250MGHYW030796
Lactisole free acid is a small-molecule antagonist of the sweet taste receptor T1R3 used to inhibit sweet taste signaling and study glucose-sensing pathways in cellular and pharmacological research. Supplied for research use only in solid and solution formats, it is commonly used in taste receptor assays and related in vitro studies.
  • Antagonist of the T1R3 sweet taste receptor.
  • Used for in vitro taste receptor and cellular signaling assays.
  • Available as solid (250 mg to 1 g) and 10 mM DMSO solution formats.
  • High purity: 99.8%.
  • Molecular formula C10H12O4, molecular weight 196.20 g·mol⁻¹.
  • For research use only; not for human consumption.