Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 2-hydroxy-, compound with dovitinib (lactate) | 692737-80-7 | MFCD23103506 | 99.8% | 482.51 g/mol | C24H27FN6O4 | 200 MG
SDP

Catalog No. 5000245540
Encompass_Preferred

Dovitinib lactate is the lactate salt of dovitinib, a multi-targeted tyrosine kinase inhibitor active at low nanomolar concentrations against FLT3, c-Kit, FGFR1/3, VEGFR1/2/3, and PDGFRα/β. Supplied as a light-yellow solid for research use, the product is provided in a 200 mg quantity and is intended for preclinical kinase and cell signaling studies.

  • Multi-targeted tyrosine kinase inhibitor with low-nanomolar IC50s against FLT3, c-Kit, FGFR1/3, VEGFR1/2/3, and PDGFRα/β.
  • Lactate salt form for improved handling and solubility.
  • Molecular formula C24H27FN6O4; molecular weight 482.51 g/mol.
  • Purity approximately 99.8% as reported by batch certificate of analysis.
  • Soluble in DMSO (~25 mg/mL); use freshly opened DMSO due to hygroscopicity.
  • Store sealed at 4°C and protect from moisture.
  • For laboratory research use only; not for human or clinical use.

Catalog No. 50-002-45540 Supplier Medchemexpress LLC Supplier No. HY10207200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dovitinib lactate is the lactate salt of dovitinib, a multi-targeted tyrosine kinase inhibitor active at low nanomolar concentrations against FLT3, c-Kit, FGFR1/3, VEGFR1/2/3, and PDGFRα/β. Supplied as a light-yellow solid for research use, the product is provided in a 200 mg quantity and is intended for preclinical kinase and cell signaling studies.

  • Multi-targeted tyrosine kinase inhibitor with low-nanomolar IC50s against FLT3, c-Kit, FGFR1/3, VEGFR1/2/3, and PDGFRα/β.
  • Lactate salt form for improved handling and solubility.
  • Molecular formula C24H27FN6O4; molecular weight 482.51 g/mol.
  • Purity approximately 99.8% as reported by batch certificate of analysis.
  • Soluble in DMSO (~25 mg/mL); use freshly opened DMSO due to hygroscopicity.
  • Store sealed at 4°C and protect from moisture.
  • For laboratory research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.