Learn More
Medchemexpress LLC Propanoic acid, 2-hydroxy-, homopolymer | 26100-51-6 | MFCD00081867 | ≥98.0% | 60,000 (average) | (C3H6O3)x | 5 G
SDP
Catalog No. 5000281395
Encompass_Preferred
Polylactic acid (PLA) is a biodegradable aliphatic polyester provided as a research-grade biochemical reagent with ≥98.0% purity and an average molecular weight of approximately 60,000 Da (CAS 26100-51-6). It is used in materials science and biomedical research as a polymeric scaffold, filler, or assay reagent.
  • Biodegradable thermoplastic polyester.
  • High purity suitable for research applications.
  • Average molecular weight ~60,000 Da for consistent properties.
  • Available in multiple pack sizes, including 5 G.
  • Useful for polymer synthesis, tissue engineering, and material testing.
Catalog No. 50-002-81395 Supplier Medchemexpress LLC Supplier No. HYW2503135G
Polylactic acid (PLA) is a biodegradable aliphatic polyester provided as a research-grade biochemical reagent with ≥98.0% purity and an average molecular weight of approximately 60,000 Da (CAS 26100-51-6). It is used in materials science and biomedical research as a polymeric scaffold, filler, or assay reagent.
  • Biodegradable thermoplastic polyester.
  • High purity suitable for research applications.
  • Average molecular weight ~60,000 Da for consistent properties.
  • Available in multiple pack sizes, including 5 G.
  • Useful for polymer synthesis, tissue engineering, and material testing.