Learn More
Medchemexpress LLC Propanoic acid, 2-hydroxy-, homopolymer | 26100-51-6 | MFCD00081867 | ≥98.0% | 60000 (Average) | (C3H6O3)x | 100 MG
SDP
Catalog No. 5000280900
Encompass_Preferred
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester commonly used in research for drug delivery systems, tissue engineering scaffolds, and biodegradable device fabrication. The material is supplied as a solid (white to off-white) with an average molecular weight of ~60,000 and is registered under CAS 26100-51-6.
  • Biodegradable and biocompatible polymer.
  • Used as a carrier for controlled-release drug delivery studies.
  • Suitable scaffold material for tissue engineering research.
  • Compatible with 3D printing and biodegradable device fabrication.
  • Available in small pack sizes for laboratory research.
  • Solid, white to off-white appearance for easy handling.
Catalog No. 50-002-80900 Supplier Medchemexpress LLC Supplier No. HYW250313100MG
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester commonly used in research for drug delivery systems, tissue engineering scaffolds, and biodegradable device fabrication. The material is supplied as a solid (white to off-white) with an average molecular weight of ~60,000 and is registered under CAS 26100-51-6.
  • Biodegradable and biocompatible polymer.
  • Used as a carrier for controlled-release drug delivery studies.
  • Suitable scaffold material for tissue engineering research.
  • Compatible with 3D printing and biodegradable device fabrication.
  • Available in small pack sizes for laboratory research.
  • Solid, white to off-white appearance for easy handling.