Learn More
Medchemexpress LLC Propanoic acid, 2-hydroxy-, sodium salt (1:1) | 72-17-3 | 60.0% | 1 ML
SDP
Catalog No. 5000410544
Encompass_Preferred
Lactic acid sodium (60% w/w in water) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. As an oncometabolite, it plays an immune protective role in anti-tumor immunity. It also possesses antimicrobial activity and can be utilized as a food preservative. This product is a 60% (w/w) solution in water, with 1 mL containing 798 mg of Lactic acid sodium at a concentration of 7.12 M.
  • Decreases cell proliferation and migration.
  • Switches cell metabolism towards oxidative phosphorylation.
  • Increases protein and mRNA expression levels of MCT1 and HCAR1.
Catalog No. 50-004-10544 Supplier Medchemexpress LLC Supplier No. HYB2227B1ML
Lactic acid sodium (60% w/w in water) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. As an oncometabolite, it plays an immune protective role in anti-tumor immunity. It also possesses antimicrobial activity and can be utilized as a food preservative. This product is a 60% (w/w) solution in water, with 1 mL containing 798 mg of Lactic acid sodium at a concentration of 7.12 M.
  • Decreases cell proliferation and migration.
  • Switches cell metabolism towards oxidative phosphorylation.
  • Increases protein and mRNA expression levels of MCT1 and HCAR1.