Learn More
Medchemexpress LLC Propanoic acid, 2-hydroxy-, sodium salt (1:1) | 72-17-3 | >98.0% | 200 UL
SDP
Catalog No. 5000410545
Encompass_Preferred
Lactic acid sodium (60% w/w in water) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. It is also an oncometabolite and has an immune protective role in anti-tumor immunity. Additionally, lactic acid sodium (60% w/w in water) possesses antimicrobial activity, making it usable as a food preservative.
  • Hydroxycarboxylic acid receptor 1 (HCAR1) activator
  • Epigenetic modulator inducing lysine residues lactylation
  • Glycolysis end-product
  • Oncometabolite
  • Immune protective role in anti-tumor immunity
  • Antimicrobial activity; can be used as a food preservative
  • Contains 60% w/w of lactic acid sodium
Catalog No. 50-004-10545 Supplier Medchemexpress LLC Supplier No. HYB2227B200UL
Lactic acid sodium (60% w/w in water) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. It is also an oncometabolite and has an immune protective role in anti-tumor immunity. Additionally, lactic acid sodium (60% w/w in water) possesses antimicrobial activity, making it usable as a food preservative.
  • Hydroxycarboxylic acid receptor 1 (HCAR1) activator
  • Epigenetic modulator inducing lysine residues lactylation
  • Glycolysis end-product
  • Oncometabolite
  • Immune protective role in anti-tumor immunity
  • Antimicrobial activity; can be used as a food preservative
  • Contains 60% w/w of lactic acid sodium