Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[[13-(4-formylphenyl)-13-oxo-3,6,9-trioxa-12-azatridec-1-yl]oxy]- | 1309460-27-2 | MFCD18916985 | 99.1% | 397.42 g/mol | C19H27NO8 | 1 G
SDP

Catalog No. 5000202266
Encompass_Preferred

A noncleavable polyethylene glycol (PEG) linker intended for antibody-drug conjugate research. It provides a 4-unit PEG spacer with an aldehyde-phenyl amido linkage and a terminal carboxylic acid for bioconjugation; supplied as a viscous, off-white to light yellow liquid with high purity and specific storage requirements.

  • Noncleavable linker for antibody-drug conjugate research.
  • High purity: 99.1%.
  • Molecular weight: 397.42 g/mol.
  • Chemical formula: C19H27NO8.
  • CAS number: 1309460-27-2.
  • Appearance: viscous liquid, off-white to light yellow.
  • Storage: store at 4°C under nitrogen; in solution, -20°C (short term) or -80°C (long term).
  • Packaged quantity: 1 G.

Catalog No. 50-002-02266 Supplier Medchemexpress LLC Supplier No. HY1300971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A noncleavable polyethylene glycol (PEG) linker intended for antibody-drug conjugate research. It provides a 4-unit PEG spacer with an aldehyde-phenyl amido linkage and a terminal carboxylic acid for bioconjugation; supplied as a viscous, off-white to light yellow liquid with high purity and specific storage requirements.

  • Noncleavable linker for antibody-drug conjugate research.
  • High purity: 99.1%.
  • Molecular weight: 397.42 g/mol.
  • Chemical formula: C19H27NO8.
  • CAS number: 1309460-27-2.
  • Appearance: viscous liquid, off-white to light yellow.
  • Storage: store at 4°C under nitrogen; in solution, -20°C (short term) or -80°C (long term).
  • Packaged quantity: 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.