Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl] oxy] | 2682112-08-7 | MFCD00869897 | 97.0% | C20H22N2O9 | 10MG
SDP
Catalog No. 5000262032
Encompass_Preferred
Thalidomide-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a PEG3 spacer terminating in a carboxylic acid. It is designed as a building block for PROTAC and targeted protein degradation research, supplied as a solid with high reported purity and DMSO solubility.
  • Incorporates cereblon-binding thalidomide scaffold for E3 recruitment.
  • PEG3 linker provides a flexible spacer for target engagement.
  • Terminal carboxylic acid enables amide coupling and conjugation chemistry.
  • High reported purity (96.95%) suitable for synthesis and analytical work.
  • Soluble in DMSO for straightforward stock solution preparation.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-62032 Supplier Medchemexpress LLC Supplier No. HY13877510MG
Thalidomide-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a PEG3 spacer terminating in a carboxylic acid. It is designed as a building block for PROTAC and targeted protein degradation research, supplied as a solid with high reported purity and DMSO solubility.
  • Incorporates cereblon-binding thalidomide scaffold for E3 recruitment.
  • PEG3 linker provides a flexible spacer for target engagement.
  • Terminal carboxylic acid enables amide coupling and conjugation chemistry.
  • High reported purity (96.95%) suitable for synthesis and analytical work.
  • Soluble in DMSO for straightforward stock solution preparation.
  • Available in small laboratory pack sizes for research use.