Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]- | 663921-15-1 | MFCD11041129 | 99.9% | 265.30 g/mol | C11H23NO6 | 100 MG
SDP

Catalog No. 5000202942
Encompass_Preferred

NH2-PEG4-CH2CH2COOH is a cleavable four-unit polyethylene glycol (PEG) linker with an N-terminal amine and a terminal propanoic acid, designed for use in antibody-drug conjugate (ADC) and PROTAC synthesis.

  • Cleavable 4-unit PEG linker.
  • Amine functional group for conjugation.
  • Terminal carboxylic acid for coupling reactions.
  • High purity (99.9%).
  • Molecular weight 265.30 g/mol.
  • Soluble in DMSO.
  • Available as a 100 MG pack.

Catalog No. 50-002-02942 Supplier Medchemexpress LLC Supplier No. HYW021787100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-PEG4-CH2CH2COOH is a cleavable four-unit polyethylene glycol (PEG) linker with an N-terminal amine and a terminal propanoic acid, designed for use in antibody-drug conjugate (ADC) and PROTAC synthesis.

  • Cleavable 4-unit PEG linker.
  • Amine functional group for conjugation.
  • Terminal carboxylic acid for coupling reactions.
  • High purity (99.9%).
  • Molecular weight 265.30 g/mol.
  • Soluble in DMSO.
  • Available as a 100 MG pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.