Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]- | 1056024-94-2 | 98.0% | 247.25 | C9H17N3O5 | 10 G
SDP
Catalog No. 5000202206
Encompass_Preferred
N3-PEG3-CH2CH2COOH is an azido-terminated PEG3 carboxylic acid linker used for click chemistry and as a PROTAC and ADC linker. It contains an azide functional group for copper-catalyzed and strain-promoted azide-alkyne conjugations, is supplied as a colorless to light yellow liquid, and is highly soluble in DMSO.
  • Azide-containing PEG3 spacer for click chemistry.
  • Compatible with CuAAC and SPAAC conjugation reactions.
  • Purity 98.0% and liquid appearance (colorless to light yellow).
  • High solubility in DMSO (100 mg/mL) and protocols for in vivo formulations.
  • Store protected from light at -20°C; in solvent store at -80°C for long term.
Catalog No. 50-002-02206 Supplier Medchemexpress LLC Supplier No. HY4249010G
N3-PEG3-CH2CH2COOH is an azido-terminated PEG3 carboxylic acid linker used for click chemistry and as a PROTAC and ADC linker. It contains an azide functional group for copper-catalyzed and strain-promoted azide-alkyne conjugations, is supplied as a colorless to light yellow liquid, and is highly soluble in DMSO.
  • Azide-containing PEG3 spacer for click chemistry.
  • Compatible with CuAAC and SPAAC conjugation reactions.
  • Purity 98.0% and liquid appearance (colorless to light yellow).
  • High solubility in DMSO (100 mg/mL) and protocols for in vivo formulations.
  • Store protected from light at -20°C; in solvent store at -80°C for long term.