Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]-, 2,5-dioxo-1-pyrrolidinyl ester | 1245718-89-1 | 99.9% | 344.32 | C13H20N4O7 | 250 MG
SDP
Catalog No. 5000204280
Encompass_Preferred
N3-PEG3-C2-NHS ester is an azide-terminated polyethylene glycol (PEG3) N-hydroxysuccinimide (NHS) ester linker used for bioconjugation and the synthesis of antibody-drug conjugates (ADCs). It reacts with primary amines via NHS ester chemistry and participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition for attachment to alkyne- or DBCO/BCN-containing partners.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • NHS ester reacts with primary amines for efficient conjugation.
  • Three-unit PEG spacer provides hydrophilicity and flexibility.
  • High purity suitable for bioconjugation workflows.
  • Supplied as a solid for convenient handling and storage.
Catalog No. 50-002-04280 Supplier Medchemexpress LLC Supplier No. HY126528250MG
N3-PEG3-C2-NHS ester is an azide-terminated polyethylene glycol (PEG3) N-hydroxysuccinimide (NHS) ester linker used for bioconjugation and the synthesis of antibody-drug conjugates (ADCs). It reacts with primary amines via NHS ester chemistry and participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition for attachment to alkyne- or DBCO/BCN-containing partners.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • NHS ester reacts with primary amines for efficient conjugation.
  • Three-unit PEG spacer provides hydrophilicity and flexibility.
  • High purity suitable for bioconjugation workflows.
  • Supplied as a solid for convenient handling and storage.