Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]-, 1,1-dimethylethyl ester | 186020-66-6 | 95.0% | 278.34 g/mol | C13H26O6 | 25 G
SDP

Catalog No. 5000202875
Encompass_Preferred

Hydroxy-PEG3-(CH2)2-Boc is a Boc-protected, hydroxy-terminated polyethylene glycol linker used as a building block in linker and PROTAC synthesis. Supplied as a white to off-white solid, it has a typical purity of 95.0% and a molecular weight of 278.34 g/mol (CAS 186020-66-6).

  • Boc-protected hydroxy-terminated PEG3 linker for synthetic chemistry.
  • Typical purity 95.0% by NMR.
  • Molecular weight 278.34 g/mol.
  • Chemical formula C13H26O6.
  • Supplied as a 25 g package for laboratory research use only.

Catalog No. 50-002-02875 Supplier Medchemexpress LLC Supplier No. HY4248825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxy-PEG3-(CH2)2-Boc is a Boc-protected, hydroxy-terminated polyethylene glycol linker used as a building block in linker and PROTAC synthesis. Supplied as a white to off-white solid, it has a typical purity of 95.0% and a molecular weight of 278.34 g/mol (CAS 186020-66-6).

  • Boc-protected hydroxy-terminated PEG3 linker for synthetic chemistry.
  • Typical purity 95.0% by NMR.
  • Molecular weight 278.34 g/mol.
  • Chemical formula C13H26O6.
  • Supplied as a 25 g package for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.